Chromium(VI) oxide ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Chromic anhydride, Chromium trioxide

216635-100G 216635-500G
BT216635-100GEA 70.53 USD
BT216635-100G BT216635-500G
Chromium(VI) oxide ≥98.0% ACS
Chromium (VI) oxide

CAS: 1333-82-0; EC No: 215-607-8; MDL No: MFCD00010952; RTECS: GB6650000 UN No: UN1463; Haz Class: 5.1 (6.1,8); Packing Group: II Crystalline; Molecular Formula: CrO3; MW: 99.99 Melting Point: 196° (decomposes) Density (g/mL): 2.7 Hygroscopic


  • RTECS: GB6650000
  • Hygroscopic
  • Crystalline

Formula: CrO₃
Melting Pt: 196 °C (dec.)
Density: 2.7 g/mL
MDL Number: MFCD00010952
CAS Number: 1333-82-0
EINECS: 215-607-8
UN: 1463
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR