Copper(II) oxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

216590-100G 216590-25G 216590-500G
BT216590-100GEA 292 USD
BT216590-100G BT216590-25G BT216590-500G
Copper(II) oxide ≥99.995% (trace metals basis)
Copper (II) oxide

CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: CuO; MW: 79.55 Melting Point: 1330° Density (g/mL): 6.4


  • RTECS: GL7900000
  • Powder

Formula: CuO
Melting Pt: 1330 °C
Density: 6.4 g/mL
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR