Trimethyl orthovalerate ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,1,1-Trimethoxypentane, Orthovaleric Acid Trimethyl Ester

216575-100G 216575-25G 216575-5G
BT216575-100GEA 375 USD
BT216575-100G BT216575-25G BT216575-5G
Trimethyl orthovalerate ≥98%
Trimethyl orthovalerate

CAS: 13820-09-2; EC No: 237-496-5; MDL No: MFCD00008481 UN No: UN1993; Haz Class: 3; Packing Group: III Liquid; Linear Formula: CH3CH2CH2CH2C(OCH3)3; Molecular Formula: C8H18O3; MW: 162.23 Boiling Point: 164-166°; Flash point: 41°C (105°F) Density (g/mL): 0.941; Refractive Index: 1.410 Moisture Sensitive


  • Refractive index: 1.410
  • Moisture Sensitive
  • Liquid

Formula: C₈H₁₈O₃
Boiling Pt: 164…166 °C
Density: 0.941 g/mL
Flash Pt: 41 °C (105 °F)
MDL Number: MFCD00008481
CAS Number: 13820-09-2
EINECS: 237-496-5
ADR: 3,III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR