8-Amino-2-naphthol ≥98%
Supplier: BeanTown Chemical
Synonyms:
1-Amino-7-naphthol
216525-1G
216525-25G
216525-5G
BT216525-1GEA
26
USD
BT216525-1G
BT216525-25G
BT216525-5G
8-Amino-2-naphthol ≥98%
8-Amino-2-naphthol
CAS: 118-46-7; EC No: 204-252-4; MDL No: MFCD00004031; RTECS: QL3331000
Powder; Molecular Formula: C10H9NO; MW: 159.18
Melting Point: 206-210°
Light Sensitive
- RTECS: QL3331000
- Light Sensitive
- Powder
Formula:
C₁₀H₉NO Melting Pt: 206…210 °C |
MDL Number:
MFCD00004031 CAS Number: 118-46-7 EINECS: 204-252-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...