4-Bromo-o-toluic acid ≥98%

Supplier: BeanTown Chemical

216460-1G 216460-25G 216460-5G
BT216460-1GEA 29 USD
BT216460-1G BT216460-25G BT216460-5G
4-Bromo-o-toluic acid ≥98%
4-Bromo-o-toluic acid

CAS: 68837-59-2; EC No: 272-437-7; MDL No: MFCD00040905 Solid; Molecular Formula: C8H7BrO2; MW: 215.04 Melting Point: 180-184°


  • Solid

Formula: C₈H₇BrO₂
Melting Pt: 180…184 °C
MDL Number: MFCD00040905
CAS Number: 68837-59-2
EINECS: 272-437-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR