Succinic acid ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: 1,4-Butanedioic acid

216325-2KG 216325-500G
BT216325-2KGEA 144.81 USD
BT216325-2KG BT216325-500G
Succinic acid ≥99.0% ACS
Succinic acid

CAS: 110-15-6; EC No: 203-740-4; MDL No: MFCD00002789; RTECS: WM4900000 Crystalline ; Linear Formula: HOOCCH2CH2COOH; Molecular Formula: C4H6O4; MW: 118.09 Melting Point: 184-186°; Boiling Point: 235°; Flash point: 206°C (402°F) Density (g/mL): 1.564


  • RTECS: WM4900000
  • Crystalline

Formula: HOOCCH₂CH₂COOH
Boiling Pt: 235 °C
Melting Pt: 184…186 °C
Density: 1.564 g/mL
Flash Pt: 206 °C (402 °F)
MDL Number: MFCD00002789
CAS Number: 110-15-6
EINECS: 203-740-4
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfd381e6e0e4cbde676a92c9e8d6aacfac635ade&vid=0b14e1b7f82b03468fc82d224a0ee894952eb200&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min
Melting range 185.0-187.0°C
Insoluble matter ≤0.01%
Residue after ignition ≤0.02%
Chloride (Cl) ≤0.001%
Phosphate (PO4) ≤0.001%
Sulfate (SO4) ≤0.003%
Nitrogen compounds (as N) ≤0.001%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤5ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR