7-Bromoindole ≥98%
Supplier: BeanTown Chemical
Synonyms:
7-Bromo-1H-indole
216320-1G
216320-250MG
216320-5G
BT216320-1GEA
292
USD
BT216320-1G
BT216320-250MG
BT216320-5G
7-Bromoindole ≥98%
7-Bromoindole
CAS: 51417-51-7; MDL No: MFCD00799492
Powder; Molecular Formula: C8H6BrN; MW: 196.04
Melting Point: 41-44°
Light Sensitive
- Light Sensitive
- Powder
Formula:
C₈H₆BrN Melting Pt: 41…44 °C |
MDL Number:
MFCD00799492 CAS Number: 51417-51-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...