Aluminium hydroxide

Supplier: BeanTown Chemical

216265-100G 216265-2KG 216265-500G
BT216265-100GEA 27.39 USD
BT216265-100G BT216265-2KG BT216265-500G
Aluminium hydroxide
Aluminium hydroxide

CAS: 21645-51-2; EC No: 244-492-7; MDL No: MFCD00003420; RTECS: BD0940000 Granular; Linear Formula: Al(OH)3; MW: 78.00


  • RTECS: BD0940000
  • Granular

Formula: Al(OH)₃
MDL Number: MFCD00003420
CAS Number: 21645-51-2
EINECS: 244-492-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR