Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥99%
Supplier: BeanTown Chemical
Synonyms:
3-Amino-4-carbethoxypyrazole, Ethyl 3-amino-1H-pyrazole-4-carboxylate
216260-100G
216260-25G
216260-5G
BT216260-100GEA
1208
USD
BT216260-100G
BT216260-25G
BT216260-5G
Ethyl-3-amino-1H-pyrazole-4-carboxylate ≥99%
Ethyl 3-amino-1H-pyrazole-4-carboxylate
CAS: 6994-25-8; EC No: 230-262-3; MDL No: MFCD00005238
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: C6H9N3O2; MW: 155.16
Melting Point: 104-107°
- Solid
Formula:
C₆H₉N₃O₂ Melting Pt: 104…107 °C |
MDL Number:
MFCD00005238 CAS Number: 6994-25-8 EINECS: 230-262-3 ADR: 9,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...