Trimethyloxonium tetrafluoroborate ≥96%
Supplier: BeanTown Chemical
216255-10G
216255-250G
216255-50G
BT216255-10GEA
78.02
USD
BT216255-10G
BT216255-250G
BT216255-50G
Trimethyloxonium tetrafluoroborate ≥96%
Trimethyloxonium tetrafluoroborate
CAS: 420-37-1; EC No: 206-994-4; MDL No: MFCD00011798
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Linear Formula: O(CH3)3BF4; Molecular Formula: C3H9BF4O; MW: 147.91
Melting Point: 190° (decomposes)
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₃H₉BF₄O Melting Pt: 190 °C (dec.) |
MDL Number:
MFCD00011798 CAS Number: 420-37-1 EINECS: 206-994-4 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...