Tin, Plasma Standard Solution, Sn 1000 µg/ml

Supplier: BeanTown Chemical

216235-500ML
BT216235-100MLEA 88.65 USD
BT216235-100ML BT216235-500ML BT216235-50ML
Tin, Plasma Standard Solution, Sn 1000 µg/ml
Standards Cationic Standards Tin Standards

EC No: 231-595-7
Haz class: 8
Packing group: II
GHS symbol: GHS05

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR