(±)-2-Methyl-3-pentanol ≥99%
Supplier: BeanTown Chemical
216165-10G
216165-50G
BT216165-10GEA
88.48
USD
BT216165-10G
BT216165-50G
(±)-2-Methyl-3-pentanol ≥99%
(±)-2-Methyl-3-pentanol
CAS: 565-67-3; EC No: 209-286-3; MDL No: MFCD00004567; RTECS: SA7400000
UN No: UN2282; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3CH2CH(OH)CH(CH3)2; Molecular Formula: C6H14O; MW: 102.17
Boiling Point: 128°; Flash point: 46°C (115°F)
Density (g/mL): 0.819; Refractive Index: 1.417
- RTECS: SA7400000
- Refractive index: 1.417
- Liquid
Formula:
C₆H₁₄O MW: 102.18 g/mol Boiling Pt: 128 Melting Pt: -53 Density: 0.819 g/mL Flash Pt: 46 °C (115 °F) |
MDL Number:
MFCD00004567 CAS Number: 565-67-3 EINECS: 209-286-3 UN: 2282 ADR: 3 III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...