Potassium hydroxide ≥85% ACS
Supplier: BeanTown Chemical
Synonyms:
Potash caustic, Potassium hydrate, Caustic potash
216145-2KG
216145-500G
BT216145-2KGEA
123.48
USD
BT216145-2KG
BT216145-500G
Potassium hydroxide ≥85% ACS
Potassium hydroxide
CAS: 1310-58-3; EC No: 215-181-3; MDL No: MFCD00003553; RTECS: TT2100000
UN No: UN1813; Haz Class: 8; Packing Group: II
Pellets; Linear Formula: KOH; MW: 56.11
Melting Point: 361°; Boiling Point: 1320°
Density (g/mL): 2.044
Air Sensitive , Hygroscopic
- RTECS: TT2100000
- Air Sensitive, Hygroscopic
- Pellets
Formula:
KOH Boiling Pt: 1320 °C Melting Pt: 361 °C Density: 2.044 g/mL |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 EINECS: 215-181-3 UN: 1813 ADR: 8,II |
Specification Test Results
Assay | 85.0% min. |
Potassium carbonate (K2CO3) | ≤2.0% |
Chloride (Cl) | ≤0.01% |
Nitrogen compounds (as N) | ≤0.001% |
Phosphate (PO4) | ≤5ppm |
Sulfate (SO4) | ≤0.003% |
Heavy metals (as Ag) | ≤0.001% |
Iron (Fe) | ≤0.001% |
Nickel (Ni) | ≤0.001% |
Calcium (Ca) | ≤0.005% |
Magnesium (Mg) | ≤0.002% |
Sodium (Na) | ≤0.05% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...