Platinum : rhodium (94:6) (w/w), wire Ø 0.5 mm for ISA type B standard grade thermocouple
Supplier: BeanTown Chemical
216100-100CM
216100-25CM
BT216100-100CMEA
831
USD
BT216100-100CM
BT216100-25CM
Platinum : rhodium (94:6) (w/w), wire Ø 0.5 mm for ISA type B standard grade thermocouple
Platinum : rhodium
MDL No: MFCD00198186
Solid; Formula: Pt:Rh; 94:6 wt%
- Solid