Sodium triacetoxyborohydride ≥95%
Supplier: BeanTown Chemical
216085-100G
216085-25G
216085-5G
BT216085-100GEA
170.45
USD
BT216085-100G
BT216085-25G
BT216085-5G
Sodium triacetoxyborohydride ≥95%
Sodium triacetoxyborohydride
CAS: 56553-60-7; MDL No: MFCD00012211
UN No: UN3396; Haz Class: 4.3 (4.1); Packing Group: III
Solid; Linear Formula: (CH3COO)3BHNa; MW: 211.94
Melting Point: 116-120°
Density (g/mL): 1.43
Moisture Sensitive
- Moisture Sensitive
- Solid
Formula:
C₆H₁₀BNaO₆ Melting Pt: 116…120 °C Density: 1.43 g/mL |
MDL Number:
MFCD00012211 CAS Number: 56553-60-7 UN: 3396 ADR: 4.3,III |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...