Sodium metaarsenite 0.1 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Sodium dioxoarsenate

216065-500ML
BT216065-500MLEA 96.55 USD
BT216065-500ML
Sodium metaarsenite 0.1 N in water volumetric solution
Sodium metaarsenite

CAS: 7784-46-5; EC No: 232-070-5; MDL No: MFCD00003472 UN No: UN1686; Haz Class: 6.1; Packing Group: II Liquid; Molecular Formula: NaAsO2; MW: 129.91


  • Liquid

Formula: NaAsO₂
MW: 129.91
Melting Pt: 615
Density: 1.87
MDL Number: MFCD00003472
CAS Number: 7784-46-5
EINECS: 232-070-5
UN: 1686
ADR: 6.1 II
Merck Index: 13,08651

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR