Nickel (II) oxide ≥99.995% (trace metals basis)
Supplier: BeanTown Chemical
216045-100G
216045-25G
BT216045-100GEA
675
USD
BT216045-100G
BT216045-25G
Nickel (II) oxide ≥99.995% (trace metals basis)
Nickel (II) oxide
CAS: 1313-99-1; EC No: 215-215-7; MDL No: MFCD00011145; RTECS: QR8400000
3-12mm Pieces; Molecular Formula: NiO ; MW: 74.71
Melting Point: 1984°
Density (g/mL): 6.67
- RTECS: QR8400000
- 3-12mm Pieces
Formula:
NiO Melting Pt: 1984 °C Density: 6.67 g/mL |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...