Cyclohexanone ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Pimelic Ketone

215915-1L 215915-4L 215915-500ML
BT215915-1LEA 51.68 USD
BT215915-1L BT215915-4L BT215915-500ML
Cyclohexanone ≥99.0% ACS
Cyclohexanone

CAS: 108-94-1; EC No: 203-631-1; MDL No: MFCD00001625; RTECS: GW1050000 UN No: UN1915; Haz Class: 3; Packing Group: III Liquid; Molecular Formula: C6H10O; MW: 98.15 Melting Point: -47°; Boiling Point: 155°; Flash point: 44°C (111°F) Density (g/mL): 0.947; Refractive Index: 1.450


  • RTECS: GW1050000
  • Refractive index: 1.450
  • Liquid

Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6
Melting Pt: -31
Density: 0.947 g/mL
Flash Pt: 44 °C (111 °F)
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3 III
Merck Index: 13,02755
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b8701dac7edb70a452483cd4f1bd716ca29ba9aa&vid=0360daffe7606174162954ba58a6ba2b78e28dfb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0% min.
Color (APHA) ≤10
Residue after evaporation ≤0.05%
Cyclohexanol ≤0.05%
Free acid (as formic acid) ≤0.01%
Water ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR