1,2,3-Trifluoro-4-nitrobenzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,3,4-Trifluoronitrobenzene

215865-100G 215865-25G 215865-5G
BT215865-100GEA 468 USD
BT215865-100G BT215865-25G BT215865-5G
1,2,3-Trifluoro-4-nitrobenzene ≥97%
1,2,3-Trifluoro-4-nitrobenzene

CAS: 771-69-7; EC No: 212-238-4; MDL No: MFCD00041546 UN No: UN2810; Haz Class: 6.1; Packing Group: III Liquid; Molecular Formula: C6H2F3NO2; MW: 177.08 Boiling Point: 92°/20 mmHg; Flash point: 93°C (199°F) Density (g/mL): 1.541; Refractive Index: 1.492


  • Refractive index: 1.492
  • Liquid

Formula: C₆H₂F₃NO₂
Boiling Pt: 92 °C/20 mmHg
Density: 1.541 g/mL
Flash Pt: 93 °C (199 °F)
MDL Number: MFCD00041546
CAS Number: 771-69-7
EINECS: 212-238-4
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR