2-(Dicyclohexylphosphino)biphenyl ≥98%

Supplier: BeanTown Chemical

Synonyms: (2-Biphenyl)dicyclohexylphosphine

215845-1G 215845-25G 215845-5G
BT215845-1GEA 41 USD
BT215845-1G BT215845-25G BT215845-5G
2-(Dicyclohexylphosphino)biphenyl ≥98%
2-(Dicyclohexylphosphino)biphenyl

CAS: 247940-06-3; EC No: 480-030-2; MDL No: MFCD01862441 Solid; Molecular Formula: C24H31P; MW: 350.48 Melting Point: 102-106° Air Sensitive


  • Air Sensitive
  • Solid

Formula: C₂₄H₃₁P
MW: 350.48 g/mol
Melting Pt: 102…106 °C
MDL Number: MFCD01862441
CAS Number: 247940-06-3
EINECS: 480-030-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR