Cobalt(II,III) oxide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Tricobalt tetraoxide, Cobaltic tetraoxide

215780-100G 215780-25G 215780-500G
BT215780-100GEA 110.63 USD
BT215780-100G BT215780-25G BT215780-500G
Cobalt(II,III) oxide ≥99.5%
Cobalt (II,III) oxide

CAS: 1308-06-1; EC No: 215-157-2; MDL No: MFCD00010939; RTECS: GG2500000 UN No: UN3077; Haz Class: 9; Packing Group: III -400 Mesh Powder; Molecular Formula: Co3O4; MW: 240.80 Melting Point: 895° (decomposes) Density (g/mL): 6.11


  • RTECS: GG2500000
  • -400 Mesh Powder

Formula: Co₃O₄
MW: 240.8 g/mol
Melting Pt: 895 (dec.)
Density: 6.11 g/mL
MDL Number: MFCD00010939
CAS Number: 1308-06-1
EINECS: 215-157-2
ADR: 9 III
Merck Index: 13,02454

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR