Indium phosphide ≥99.99%, polycrystalline lumps 2-8 mm (0.08-0.3 in)

Supplier: BeanTown Chemical

Synonyms: Indium(III) phosphide, Indium phosphide

215775-1G 215775-25G 215775-5G
BT215775-1GEA 92 USD
BT215775-1G BT215775-25G BT215775-5G
Indium phosphide ≥99.99%, polycrystalline lumps 2-8 mm (0.08-0.3 in)
Indium phosphide

CAS: 22398-80-7; EC No: 244-959-5; MDL No: MFCD00016153; RTECS: NL1800000 Pieces; Molecular Formula: InP; MW: 145.79


  • RTECS: NL1800000
  • Pieces

Formula: InP
MDL Number: MFCD00016153
CAS Number: 22398-80-7
EINECS: 244-959-5

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR