Copper(II) bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: Cupric bromide, Cuprous bromide

215750-100G 215750-25G 215750-500G
BT215750-100GEA 54.78 USD
BT215750-100G BT215750-25G BT215750-500G
Copper(II) bromide ≥99%
Copper (II) bromide

CAS: 7789-45-9; EC No: 232-167-2; MDL No: MFCD00010970 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: CuBr2; MW: 223.36 Melting Point: 498° Density (g/mL): 4.77 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: Br₂Cu
MW: 223.35 g/mol
Boiling Pt: 900
Melting Pt: 498
Density: 4.77 g/mL
MDL Number: MFCD00010970
CAS Number: 7789-45-9
EINECS: 232-167-2
UN: 3260
ADR: 8 III
Merck Index: 13,02656

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR