4-Nitrophthalamide ≥98%
Supplier: BeanTown Chemical
215745-1G
215745-25G
215745-5G
BT215745-1GEA
33
USD
BT215745-1G
BT215745-25G
BT215745-5G
4-Nitrophthalamide ≥98%
4-Nitrophthalamide
CAS: 13138-53-9; EC No: 236-070-6; MDL No: MFCD00052702
Powder; Molecular Formula: C8H7N3O4 ; MW: 209.16
Melting Point: 195° (decomposes)
- Powder
Formula:
C₈H₇N₃O₄ Melting Pt: 195 °C (dec.) |
MDL Number:
MFCD00052702 CAS Number: 13138-53-9 EINECS: 236-070-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...