2,2'-O-Cyclocytidine hydrochloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride

215600-1G 215600-5G
BT215600-1GEA 42 USD
BT215600-1G BT215600-5G
2,2'-O-Cyclocytidine hydrochloride ≥98%
2,2'-O-Cyclocytidine hydrochloride

CAS: 10212-25-6; EC No: 233-515-6; MDL No: MFCD00012636; RTECS: LV2615000 Crystalline/Powder; Molecular Formula: C9H12ClN3O4; MW: 261.66


  • RTECS: LV2615000
  • Crystalline/Powder

Formula: C₉H₁₁N₃O₄
MW: 261.66 g/mol
MDL Number: MFCD00012636
CAS Number: 10212-25-6
EINECS: 233-515-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR