≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

215585-1KG 215585-250G 215585-5KG
BT215585-1KGEA 202.16 USD
BT215585-1KG BT215585-250G BT215585-5KG
≥98.0% ACS
di-Potassium hydrogen phosphate

CAS: 7758-11-4; EC No: 231-834-5; MDL No: MFCD00011383; RTECS: TC5580000 Granular; Linear Formula: K2HPO4; MW: 174.18 Melting Point: 340° Density (g/mL): 2.3 Hygroscopic


  • RTECS: TC5580000
  • Hygroscopic
  • Granular

Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Density: 2.3 g/mL
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0% min.
pH of 5% solution at 25°C 8.5-9.6
Insoluble matter ≤0.01%
Loss on drying (105°C) ≤1.0%
Chloride (Cl) ≤0.003%
Nitrogen compounds (as N) ≤0.001%
Sulfate (SO4) ≤0.005%
Heavy metals (as Pb) ≤5ppm
Iron (Fe) ≤0.001%
Sodium (Na) ≤0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR