Copper(II) oxide ≥99.9995% (trace metals basis)
Supplier: BeanTown Chemical
215580-100G
215580-25G
215580-5G
BT215580-100GEA
656
USD
BT215580-100G
BT215580-25G
BT215580-5G
Copper(II) oxide ≥99.9995% (trace metals basis)
Copper (II) oxide
CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: CuO; MW: 79.55
Melting Point: 1330°
Density (g/mL): 6.4
- RTECS: GL7900000
- Powder
Formula:
CuO Melting Pt: 1330 °C Density: 6.4 g/mL |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 99.9995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...