Copper(II) oxide ≥99.9995%

Supplier: BeanTown Chemical

Synonyms: Cupric oxide, Copper oxide, Copper monoxide granulated

215580-100G 215580-25G 215580-5G
BT215580-100GEA 656 USD
BT215580-100G BT215580-25G BT215580-5G
Copper(II) oxide ≥99.9995%
Copper (II) oxide

CAS: 1317-38-0; EC No: 215-269-1; MDL No: MFCD00010979; RTECS: GL7900000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: CuO; MW: 79.55 Melting Point: 1330° Density (g/mL): 6.4


  • RTECS: GL7900000
  • Powder

Formula: CuO
MW: 79.55 g/mol
Boiling Pt: 2000
Melting Pt: 1326
Density: 6.4 g/mL
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9 III
Merck Index: 13,02674

Order Now

Specification Test Results

Assay 99.9995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR