Ammonium tetrafluoroborate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Ammonium fluoroborate, ammonium fluoborate ammonium borofluoride ammonium fluoroborate [qr] ammonium tetrafluoroborate
215545-2KG
215545-500G
BT215545-2KGEA
172
USD
BT215545-2KG
BT215545-500G
Ammonium tetrafluoroborate ≥97%
Ammonium tetrafluoroborate
CAS: 13826-83-0; EC No: 237-531-4; MDL No: MFCD06245785
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: NH4BF4 ; MW: 104.84
Melting Point: 230°
Density (g/mL): 1.871
Hygroscopic
- Hygroscopic
- Powder
Formula:
H₄BF₄N Melting Pt: 230 °C Density: 1.871 g/mL |
MDL Number:
MFCD06245785 CAS Number: 13826-83-0 EINECS: 237-531-4 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...