Crystal violet ≥90% ACS
Supplier: BeanTown Chemical
215520-100G
215520-25G
BT215520-100GEA
80
USD
BT215520-100G
BT215520-25G
Crystal violet ≥90% ACS
Crystal Violet
CAS: 548-62-9; EC No: 208-953-6; MDL No: MFCD00011750; RTECS: BO9000000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: C25H30ClN3; MW: 407.99
Melting Point: 205° (decomposes)
- RTECS: BO9000000
- Powder
Formula:
C₂₅H₃₀ClN₃ Melting Pt: 205 °C (dec.) |
MDL Number:
MFCD00011750 CAS Number: 548-62-9 EINECS: 208-953-6 UN: 3077 ADR: 9,III |
Specification Test Results
Assay | 90.0% min. |
Loss on drying | ≤7.5% |
Visual transition pH | 0.8-2.6, green-blue-violet |
Sensitivity | Passes test |
Absorbance characteristics | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...