Iron(II) perchlorate hydrate
Supplier: BeanTown Chemical
215365-100G
215365-500G
BT215365-100GEA
52.53
USD
BT215365-100G
BT215365-500G
Iron(II) perchlorate hydrate
Iron(II) perchlorate hydrate
CAS: 335159-18-7; EC No: 237-704-4; MDL No: MFCD00150234
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline ; Linear Formula: Fe(ClO4)2·xH2O; MW: 254.75 (anhydrous)
Hygroscopic
- Hygroscopic
- Crystalline
Formula:
Fe(ClO₄)₂·xH₂O MW: 254.75 g/mol (anhy) |
MDL Number:
MFCD00150234 CAS Number: 335159-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...