Deuterium chloride 20% (w/w) in D₂O (99.5% D) (Isotopic)

Supplier: BeanTown Chemical

215355-50G
BT215355-50GEA 170 USD
BT215355-50G
Deuterium chloride 20% (w/w) in D₂O (99.5% D) (Isotopic)
Deuterium chloride

CAS: 7698-05-7; EC No: 231-715-8; MDL No: MFCD00044642 UN No: UN1789; Haz Class: 8; Packing Group: II Liquid, packaged in glass ampules; Molecular Formula: DCl ; MW: 37.47 Density (g/mL): 1.2 Moisture Sensitive


  • Moisture Sensitive
  • Liquid, packaged in glass ampules

Formula: DCl
Density: 1.2 g/mL
MDL Number: MFCD00044642
CAS Number: 7698-05-7
EINECS: 231-715-8
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR