Cetrimonium chloride ≥96%
Supplier: BeanTown Chemical
|
Danger
|
Synonyms:
Cetyltrimethylammonium chloride, (1-Hexadecyl)trimethylammonium chloride, Hexadecyltrimethylammonium chloride, N-Cetyl-N,N,N-trimethylammonium chloride, N,N,N-Trimethylhexadecan-1-aminium chloride, CTAC
215345-100G
215345-25G
215345-5G
BT215345-100GEA
274.61
USD
BT215345-100G
BT215345-25G
BT215345-5G
Cetrimonium chloride ≥96%
Cetrimonium chloride
CAS: 112-02-7; EC No: 203-928-6; MDL No: MFCD00011773; RTECS: ML9145000
UN No: UN1759; Haz Class: 8; Packing Group: III
Powder; Linear Formula: CH3(CH2)15N(Cl)(CH3)3; Molecular Formula: C19H42ClN; MW: 320.00
Melting Point: 232-237°
Density (g/mL): 0.968; Refractive Index: 1.378
- RTECS: ML9145000
- Refractive index: 1.378
- Powder
Formula:
C₁₉H₄₂ClN Melting Pt: 232…237 °C Density: 0.968 g/mL |
MDL Number:
MFCD00011773 CAS Number: 112-02-7 EINECS: 203-928-6 UN: 1759 ADR: 8,III |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...