Octadecylphosphonic acid ≥97%
Supplier: BeanTown Chemical
Synonyms:
n-Octadecylphosphonic acid, ODPA
215215-1G
215215-5G
BT215215-1GEA
70
USD
BT215215-1G
BT215215-5G
Octadecylphosphonic acid ≥97%
Octadecylphosphonic acid
CAS: 4724-47-4; EC No: 225-216-4; MDL No: MFCD00015838
Crystalline; Linear Formula: CH3(CH2)17P(O)(OH)2; Molecular Formula: C18H39O3P; MW: 334.48
Melting Point: 95-100°
- Crystalline
Formula:
C₁₈H₃₉O₃P Melting Pt: 95…100 °C |
MDL Number:
MFCD00015838 CAS Number: 4724-47-4 EINECS: 225-216-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...