Nickel (II) oxide, green
Supplier: BeanTown Chemical
215110-1KG
215110-250G
BT215110-1KGEA
248.43
USD
BT215110-1KG
BT215110-250G
Nickel (II) oxide, green
Nickel (II) oxide
CAS: 1313-99-1; EC No: 215-215-7; MDL No: MFCD00011145; RTECS: QR8400000
-400 Mesh Powder; Molecular Formula: NiO; MW: 74.71
Melting Point: 1984°
Density (g/mL): 6.67
- RTECS: QR8400000
- -400 Mesh Powder
Formula:
NiO Melting Pt: 1984 °C Density: 6.67 g/mL |
MDL Number:
MFCD00011145 CAS Number: 1313-99-1 EINECS: 215-215-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...