Silver ≥99.9% (trace metals basis), spherical powder APS 1.3-3.2 µm

Supplier: BeanTown Chemical

215085-10G 215085-250G 215085-50G
BT215085-10GEA 58 USD
BT215085-10G BT215085-250G BT215085-50G
Silver ≥99.9% (trace metals basis), spherical powder APS 1.3-3.2 µm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397 UN No: UN3077; Haz Class: 9; Packing Group: III Powder, S.A. 0.3-0.7m 2/g; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • Powder, S.A. 0.3-0.7m 2/g

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
ADR: 9 III
Merck Index: 12,08647

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR