Cytidine-5'-monophosphate (synthetic) ≥99%

Supplier: BeanTown Chemical

Synonyms: 5'-Cytidylic Acid, 5'-CMP, ((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate

215080-1G 215080-500MG 215080-5G
BT215080-1GEA 71.89 USD
BT215080-1G BT215080-500MG BT215080-5G
Cytidine-5'-monophosphate (synthetic) ≥99%
Cytidine-5'-monophosphate

CAS: 63-37-6; EC No: 200-556-6; MDL No: MFCD00006544 Crystalline; Molecular Formula: C9H14N3O8P; MW: 323.20 Melting Point: 222° (decomposes)


  • Crystalline

Formula: C₉H₁₄N₃O₈P
Melting Pt: 222 °C (dec.)
MDL Number: MFCD00006544
CAS Number: 63-37-6
EINECS: 200-556-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR