Cytidine-5'-monophosphate (synthetic) ≥99%
Supplier: BeanTown Chemical
Synonyms:
5'-Cytidylic Acid, 5'-CMP, ((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl dihydrogen phosphate
215080-1G
215080-500MG
215080-5G
BT215080-1GEA
71.89
USD
BT215080-1G
BT215080-500MG
BT215080-5G
Cytidine-5'-monophosphate (synthetic) ≥99%
Cytidine-5'-monophosphate
CAS: 63-37-6; EC No: 200-556-6; MDL No: MFCD00006544
Crystalline; Molecular Formula: C9H14N3O8P; MW: 323.20
Melting Point: 222° (decomposes)
- Crystalline
Formula:
C₉H₁₄N₃O₈P Melting Pt: 222 °C (dec.) |
MDL Number:
MFCD00006544 CAS Number: 63-37-6 EINECS: 200-556-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...