Vanadium ≥99.5%, turnings ≤25 mm
Supplier: BeanTown Chemical
215055-10G
215055-250G
215055-50G
BT215055-10GEA
50.33
USD
BT215055-10G
BT215055-250G
BT215055-50G
Vanadium ≥99.5%, turnings ≤25 mm
Vanadium
CAS: 7440-62-2; EC No: 231-171-1; MDL No: MFCD00011453
Solid; Molecular Formula: V; MW: 50.94
Melting Point: 1890°; Boiling Point: 3380°
Density (g/mL): 6.11
- Solid
Formula:
V Boiling Pt: 3380 °C Melting Pt: 1890 °C Density: 6.11 g/mL |
MDL Number:
MFCD00011453 CAS Number: 7440-62-2 EINECS: 231-171-1 |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...