3-(2-Hydroxyethyl)indole ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol

215025-1G 215025-5G
BT215025-1GEA 29 USD
BT215025-1G BT215025-5G
3-(2-Hydroxyethyl)indole ≥97%
3-(2-Hydroxyethyl)indole

CAS: 526-55-6; EC No: 208-393-2; MDL No: MFCD00005659; RTECS: KL3685000 Solid; Molecular Formula: C10H11NO ; MW: 161.20 Melting Point: 56-59°; Flash point: 255°C (491°F)


  • RTECS: KL3685000
  • Solid

Formula: C₁₀H₁₁NO
MW: 161.2 g/mol
Boiling Pt: 358
Melting Pt: 56…59 °C
Flash Pt: 255 °C (491 °F)
MDL Number: MFCD00005659
CAS Number: 526-55-6
EINECS: 208-393-2
Merck Index: 12,09930

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR