N,N-Dimethylglycine ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-(Dimethylamino)acetic acid, (Dimethylamino)acetic acid
214975-100G
214975-25G
214975-5G
BT214975-100GEA
710
USD
BT214975-100G
BT214975-25G
BT214975-5G
N,N-Dimethylglycine ≥98%
N,N-Dimethylglycine
CAS: 1118-68-9; EC No: 214-267-8; MDL No: MFCD00004283; RTECS: MB9865000
Crystalline/Powder; Linear Formula: (CH3)2NCH2COOH; Molecular Formula: C4H9NO2; MW: 103.12
Melting Point: 178-182°
Hygroscopic
- RTECS: MB9865000
- Hygroscopic
- Crystalline/Powder
Formula:
C₄H₉NO₂ Melting Pt: 178…182 °C |
MDL Number:
MFCD00004283 CAS Number: 1118-68-9 EINECS: 214-267-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...