cis-4-Octene ≥97%

Supplier: BeanTown Chemical

Synonyms: (Z)-4-Octene

214790-1G 214790-5G
BT214790-1GEA 26 USD
BT214790-1G BT214790-5G
cis-4-Octene ≥97%
cis-4-Octene

CAS: 7642-15-1; EC No: 231-578-4; MDL No: MFCD00067484 UN No: UN3295; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2CH2CH=CHCH2CH2CH3; Molecular Formula: C8H16; MW: 112.22


  • Liquid

Formula: C₈H₁₆
MW: 112.22 g/mol
Boiling Pt: 123
Melting Pt: -118.7
MDL Number: MFCD00067484
CAS Number: 7642-15-1
EINECS: 231-578-4
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR