Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥96%
Supplier: BeanTown Chemical
Synonyms:
(Chloro)(perfluorophenyl)dimethylsilane
214780-1G
214780-5G
BT214780-1GEA
39
USD
BT214780-1G
BT214780-5G
Chlorodimethyl(pentafluorophenyl)silane (flophemesyl chloride) ≥96%
Chlorodimethyl(pentafluorophenyl)silane (Flophemesyl chloride)
CAS: 20082-71-7; EC No: 243-506-9; MDL No: MFCD00000500
UN No: UN2987; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C8H6ClF5Si; MW: 260.66
Boiling Point: 88-90°/10 mmHg; Flash point: 95°C (203°F)
Density (g/mL): 1.384; Refractive Index: 1.448
Moisture Sensitive
- Refractive index: 1.448
- Moisture Sensitive
- Liquid
Formula:
C₈H₆ClF₅Si Boiling Pt: 88…90 °C/10 mmHg Density: 1.384 g/mL Flash Pt: 95 °C (203 °F) |
MDL Number:
MFCD00000500 CAS Number: 20082-71-7 EINECS: 243-506-9 UN: 2987 ADR: 8,II |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...