Chromium ≥99.99% (trace metals basis), powder -60 mesh
Supplier: BeanTown Chemical
214720-100G
214720-500G
BT214720-100GEA
152.04
USD
BT214720-100G
BT214720-500G
Chromium ≥99.99% (trace metals basis), powder -60 mesh
Chromium
CAS: 7440-47-3; EC No: 231-157-5; MDL No: MFCD00010944; RTECS: GB4200000
Powder; Molecular Formula: Cr; MW: 52.00
Melting Point: 1857°; Boiling Point: 2672°
Density (g/mL): 7.14
- RTECS: GB4200000
- Powder
Formula:
Cr Boiling Pt: 2672 °C Melting Pt: 1857 °C Density: 7.14 g/mL |
MDL Number:
MFCD00010944 CAS Number: 7440-47-3 EINECS: 231-157-5 UN: 3089 ADR: 4.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...