1,4-Dibromonaphthalene ≥98%

Supplier: BeanTown Chemical

214685-25G 214685-5G
BT214685-25GEA 552 USD
BT214685-25G BT214685-5G
1,4-Dibromonaphthalene ≥98%
1,4-Dibromonaphthalene

CAS: 83-53-4; EC No: 201-484-8; MDL No: MFCD00041823 Solid; Molecular Formula: C10H6Br2; MW: 285.97


  • Solid

Formula: C₁₀H₆Br₂
MDL Number: MFCD00041823
CAS Number: 83-53-4
EINECS: 201-484-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR