Praseodymium ≥99.9% (trace rare earth metals basis), powder -40 mesh

Supplier: BeanTown Chemical


Ampuled under argon

214670-5G
BT214670-5GEA 120 USD
BT214670-5G
Praseodymium ≥99.9% (trace rare earth metals basis), powder -40 mesh
Praseodymium

CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174 UN No: UN3089; Haz Class: 4.1; Packing Group: II Powder; Molecular Formula: Pr; MW: 140.91 Melting Point: 931°; Boiling Point: 3520° Density (g/mL): 6.71


  • Powder

Formula: Pr
MW: 140.91
Boiling Pt: 3520 °C
Melting Pt: 931 °C
Density: 6.71 g/mL
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 3089
ADR: 4.1 II

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR