Tetrabutylammonium hydrogen sulfate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulfate, Tetrabutylammonium bisulfate, Tetrabutylammonium hydrogensulphate
214530-1KG
214530-250G
214530-50G
BT214530-1KGEA
309
USD
BT214530-1KG
BT214530-250G
BT214530-50G
Tetrabutylammonium hydrogen sulfate ≥97%
Tetrabutylammonium hydrogen sulphate
CAS: 32503-27-8; EC No: 251-068-5; MDL No: MFCD00011637
Solid; Linear Formula: [CH3(CH2)3]4N(HSO4); Molecular Formula: C16H37NO4S; MW: 339.54
Melting Point: 169-171°
Density (g/mL): 1.01
Hygroscopic
- Hygroscopic
- Solid
Formula:
(C₄H₉)₄NHSO₄ Melting Pt: 169…171 °C Density: 1.01 g/mL |
MDL Number:
MFCD00011637 CAS Number: 32503-27-8 EINECS: 251-068-5 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...