Magnesium chloride, anhydrous ≥99%
Supplier: BeanTown Chemical
214515-1KG
214515-5KG
BT214515-1KGEA
48.25
USD
BT214515-1KG
BT214515-5KG
Magnesium chloride, anhydrous ≥99%
Magnesium chloride
CAS: 7786-30-3; EC No: 232-094-6; MDL No: MFCD00011106; RTECS: OM2800000
Crystalline; Linear Formula: MgCl2; MW: 95.22
Melting Point: 714°
Density (g/mL): 2.32
Hygroscopic
- RTECS: OM2800000
- Hygroscopic
- Crystalline
Formula:
MgCl₂ Melting Pt: 714 °C Density: 2.32 g/mL |
MDL Number:
MFCD00011106 CAS Number: 7786-30-3 EINECS: 232-094-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...