Selenium amorphous ≥99.999% (trace metals basis), shot 2-6 mm
Supplier: BeanTown Chemical
214490-1KG
214490-250G
214490-50G
BT214490-1KGEA
778
USD
BT214490-1KG
BT214490-250G
BT214490-50G
Selenium amorphous ≥99.999% (trace metals basis), shot 2-6 mm
Selenium
CAS: 7782-49-2; EC No: 231-957-4; MDL No: MFCD00134090; RTECS: VS7700000
UN No: UN3077; Haz Class: 9; Packing Group: III
Solid; Molecular Formula: Se; MW: 78.96
Melting Point: 217°; Boiling Point: 684.9°
Density (g/mL): 4.81
- RTECS: VS7700000
- Solid
Formula:
Se Boiling Pt: 684.9 °C Melting Pt: 217 °C Density: 4.81 g/mL |
MDL Number:
MFCD00134090 CAS Number: 7782-49-2 EINECS: 231-957-4 UN: 3077 ADR: 6.1,III |
Specification Test Results
Assay | 99.999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...