Tetrahydrofuran-[D₈] ≥99.5% (Isotopic)

Supplier: BeanTown Chemical

Synonyms: Deuterated Tetrahydrofuran, Octadeuterotetrahydrofuran, Tetrahydrofuran-[²H₈], THF-d8, THF-D8, Tetrahydrofuran-d8

214460-1G
BT214460-1GEA 74.38 USD
BT214460-1G
Tetrahydrofuran-[D₈] ≥99.5% (Isotopic)
Tetrahydrofuran-[D8]

CAS: 1693-74-9; EC No: 216-898-4; MDL No: MFCD00044238 UN No: UN2056; Haz Class: 3; Packing Group: II Liquid; Molecular Formula: C4D8O; MW: 80.16 Melting Point: -106°; Boiling Point: 65-66°; Flash point: -17°C (1°F) Density (g/mL): 0.985; Refractive Index: 1.403


  • Refractive index: 1.403
  • Liquid

Formula: C₄D₈O
MW: 80.04 g/mol
Boiling Pt: 65…66 °C
Melting Pt: –106 °C
Density: 0.985 g/mL
Flash Pt: –17 °C (1 °F)
MDL Number: MFCD00044238
CAS Number: 1693-74-9
EINECS: 216-898-4
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR